3’-Bromo-5’-fluoro-2’-hydroxyacetophenone
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3-Bromo-5-fluoro-2-hydroxyphenyl)ethanone
PC27937-1G PC27937-250MG PC27937-5G
APOSPC27937-1GEA 377 EUR
APOSPC27937-1G APOSPC27937-250MG APOSPC27937-5G
3’-Bromo-5’-fluoro-2’-hydroxyacetophenone
3’-Bromo-5’-fluoro-2’-hydroxyacetophenone
Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD16742831
CAS Number: 393-62-4