1-[2-(4-Fluorophenyl)-4-methyl-1,3-thiazol-5-yl]ethan-1-one 95%
Supplier: Apollo Scientific
PC28057-1G
PC28057-5G
APOSPC28057-1GEA
180
EUR
APOSPC28057-1G
APOSPC28057-5G
1-[2-(4-Fluorophenyl)-4-methyl-1,3-thiazol-5-yl]ethan-1-one 95%
1-[2-(4-Fluorophenyl)-4-methyl-1,3-thiazol-5-yl]ethan-1-one
Formula:
C₁₂H₁₀FNOS MW: 235.277 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09295390 CAS Number: 923701-97-7 |