3-[4-(Trifluoromethyl)phenyl]-1,2-oxazol-5-amine 95%
Supplier: Apollo Scientific
PC28229-1G PC28229-5G
APOSPC28229-1GEA 224 EUR
APOSPC28229-1G APOSPC28229-5G
3-[4-(Trifluoromethyl)phenyl]-1,2-oxazol-5-amine 95%
3-[4-(Trifluoromethyl)phenyl]-1,2-oxazol-5-amine
Formula: C₁₀H₇F₃N₂O
MW: 228.171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06199355
CAS Number: 81465-84-1