5-Fluoropyrimidine-2,4-diamine
Supplier: Apollo Scientific
PC28236-100MG PC28236-1G
APOSPC28236-100MGEA 219 EUR
APOSPC28236-100MG APOSPC28236-1G
5-Fluoropyrimidine-2,4-diamine
5-Fluoropyrimidine-2,4-diamine
Formula: C₄H₅FN₄
MW: 128.108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18377712
CAS Number: 155-11-3