2,6-Difluoro-4-methoxybenzenesulphonamide
Supplier: Apollo Scientific
Warning
PC28255-1G PC28255-250MG
APOSPC28255-1GEA 134 EUR
APOSPC28255-1G APOSPC28255-250MG
2,6-Difluoro-4-methoxybenzenesulphonamide
2,6-Difluoro-4-methoxybenzenesulphonamide
Formula: C₇H₇F₂NO₃S
MW: 223.197 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18394894
CAS Number: 1261738-62-8