6-(Trifluoromethoxy)-2-oxo-2H-chromene-3-carboxylic acid
Supplier: Apollo Scientific
Warning
PC28257-1G PC28257-250MG
APOSPC28257-1GEA 168 EUR
APOSPC28257-1G APOSPC28257-250MG
6-(Trifluoromethoxy)-2-oxo-2H-chromene-3-carboxylic acid
6-(Trifluoromethoxy)-2-oxo-2H-chromene-3-carboxylic acid
Formula: C₁₁H₅F₃O₅
MW: 274.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31793556
CAS Number: 165685-08-5