2-(6-Oxo-4-(trifluoromethyl)pyrimidin-1(6H)-yl)acetonitrile
Supplier: Apollo Scientific
Warning
PC28266-1G PC28266-250MG
APOSPC28266-1GEA 685 EUR
APOSPC28266-1G APOSPC28266-250MG
2-(6-Oxo-4-(trifluoromethyl)pyrimidin-1(6H)-yl)acetonitrile
2-(6-Oxo-4-(trifluoromethyl)pyrimidin-1(6H)-yl)acetonitrile
Formula: C₇H₄F₃N₃O
MW: 203.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29042744
CAS Number: 1823188-13-1
UN: 3439
ADR: 6.1,III