6-(Trifluoromethoxy)spiro[benzo[e][1,3]oxazine-2,4'-piperidin]-4(3H)-one hydrochloride
Supplier: Apollo Scientific
Warning
PC28269-1G PC28269-250MG
APOSPC28269-1GEA 522 EUR
APOSPC28269-1G APOSPC28269-250MG
6-(Trifluoromethoxy)spiro[benzo[e][1,3]oxazine-2,4'-piperidin]-4(3H)-one hydrochloride
6-(Trifluoromethoxy)spiro[benzo[e][1,3]oxazine-2,4'-piperidin]-4(3H)-one hydrochloride
Formula: C₁₃H₁₃F₃N₂O₃·ClH
MW: 338.71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29042696
CAS Number: 1823184-48-0