7-Fluoro-2H-benzo[e][1,2,4]thiadiazin-3(4H)-one 1,1-dioxide
Supplier: Apollo Scientific
Warning
PC28271-1G PC28271-250MG
APOSPC28271-1GEA 1700 EUR
APOSPC28271-1G APOSPC28271-250MG
7-Fluoro-2H-benzo[e][1,2,4]thiadiazin-3(4H)-one 1,1-dioxide
7-Fluoro-2H-benzo[e][1,2,4]thiadiazin-3(4H)-one 1,1-dioxide
Formula: C₇H₅FN₂O₃S
MW: 216.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546146
CAS Number: 152721-97-6