8-Chloro-3-(chlorodifluoromethyl)-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Supplier: Apollo Scientific
Warning
PC28276-1G PC28276-250MG
APOSPC28276-1GEA 1360 EUR
APOSPC28276-1G APOSPC28276-250MG
8-Chloro-3-(chlorodifluoromethyl)-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
8-Chloro-3-(chlorodifluoromethyl)-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Formula: C₈H₂Cl₂F₅N₃
MW: 306.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02187042
CAS Number: 478248-86-1