1-(8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridin-3-yl)-2,2,2-trifluoroethanone
Supplier: Apollo Scientific
Warning
PC28278-1G PC28278-250MG PC28278-5G
APOSPC28278-1GEA 432 EUR
APOSPC28278-1G APOSPC28278-250MG APOSPC28278-5G
1-(8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridin-3-yl)-2,2,2-trifluoroethanone
1-(8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridin-3-yl)-2,2,2-trifluoroethanone
Formula: C₁₀H₃ClF₆N₂O
MW: 316.587 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12406877
CAS Number: 1823182-59-7