3-(Trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyrazin-8(7H)-one
Supplier: Apollo Scientific
Warning
PC28298-1G PC28298-250MG
APOSPC28298-1GEA 68.3 EUR
APOSPC28298-1G APOSPC28298-250MG
3-(Trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyrazin-8(7H)-one
3-(Trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyrazin-8(7H)-one
Formula: C₆H₃F₃N₄O
Storage Temperature: Ambient
MDL Number: MFCD31812105
CAS Number: 140911-30-4