3',5'-Difluoro-4'-methoxypropiophenone 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Difluoro-4-propanoylanisole, 1-(3,5-Difluoro-4-methoxyphenyl)propan-1-one
PC2848-1G PC2848-250MG
APOSPC2848-1GEA 321 EUR
APOSPC2848-1G APOSPC2848-250MG
3',5'-Difluoro-4'-methoxypropiophenone 97%
3',5'-Difluoro-4'-methoxypropiophenone
Formula: C₁₀H₁₀F₂O₂
MW: 200.182 g/mol
Boiling Pt: 203…204 °C
Melting Pt: 44478 °C
Density: 1.451 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00042509
CAS Number: 71292-82-5