3,5-Difluoro-4-iodoaniline 97%
Supplier: Apollo Scientific
Warning
PC2906-25G PC2906-5G
APOSPC2906-25GEA 121 EUR
APOSPC2906-25G APOSPC2906-5G
3,5-Difluoro-4-iodoaniline 97%
3,5-Difluoro-4-iodoaniline
Formula: C₆H₄F₂IN
MW: 255.004 g/mol
Melting Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07774190
CAS Number: 1542-34-3