4-Fluoro-1H-benzimidazole 98%
Supplier: Apollo Scientific
Warning
PC2936-1G PC2936-5G
APOSPC2936-1GEA 128 EUR
APOSPC2936-1G APOSPC2936-5G
4-Fluoro-1H-benzimidazole 98%
4-Fluoro-1H-benzimidazole
Formula: C₇H₅FN₂
MW: 136.126 g/mol
Melting Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD08729272
CAS Number: 5847-89-2