4-(Trifluoromethyl)-1H-benzimidazole
Supplier: Apollo Scientific
Warning
Synonyms: 7-(Trifluoromethyl)-1H-benzo[d]imidazole, 7-(Trifluoromethyl)-1H-benzimidazole
PC2937-1G PC2937-5G
APOSPC2937-1GEA 232 EUR
APOSPC2937-1G APOSPC2937-5G
4-(Trifluoromethyl)-1H-benzimidazole
4-(Trifluoromethyl)-1H-benzimidazole
Formula: C₈H₅F₃N₂
MW: 186.134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08741394
CAS Number: 392-11-0