2-Thio-5-(trifluoromethyl)-1,3-benzoxazole
Supplier: Apollo Scientific
Warning
Synonyms: 5-(Trifluoromethyl)-1,3-benzoxazole-2-thiol, 2-Sulphanyl-5-(trifluoromethyl)benzo[d]oxazole
PC300517-100MG PC300517-1G PC300517-250MG
APOSPC300517-100MGEA 0 EUR
APOSPC300517-100MG APOSPC300517-1G APOSPC300517-250MG
2-Thio-5-(trifluoromethyl)-1,3-benzoxazole
2-Thio-5-(trifluoromethyl)-1,3-benzoxazole
Formula: C₈H₄F₃NOS
MW: 219.184 g/mol
Melting Pt: 166…168 °C
Storage Temperature: Ambient
MDL Number: MFCD23183903
CAS Number: 13451-80-4