Ethyl 3-bromo-6-fluoroimidazo[1,2-a]pyridine-2-carboxylate
Supplier: Apollo Scientific
Warning
PC300532-1G PC300532-250MG
APOSPC300532-1GEA 284 EUR
APOSPC300532-1G APOSPC300532-250MG
Ethyl 3-bromo-6-fluoroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 3-bromo-6-fluoroimidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₀H₈BrFN₂O₂
MW: 287.085 g/mol
Melting Pt: 52…55 °C
Storage Temperature: Ambient
MDL Number: MFCD23159158
CAS Number: 1427460-92-1