3-Chloro-2-phenoxy-5-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
PC300538-1G PC300538-5G
APOSPC300538-1GEA 92.8 EUR
APOSPC300538-1G APOSPC300538-5G
3-Chloro-2-phenoxy-5-(trifluoromethyl)pyridine
3-Chloro-2-phenoxy-5-(trifluoromethyl)pyridine
Formula: C₁₂H₇ClF₃NO
MW: 273.638 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00244669
CAS Number: 95711-04-9