4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzhydrazide
Supplier: Apollo Scientific
Synonyms: 4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzohydrazide
PC300539-1G PC300539-5G
APOSPC300539-1GEA 193 EUR
APOSPC300539-1G APOSPC300539-5G
4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzhydrazide
4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzhydrazide
Formula: C₁₃H₉ClF₃N₃O₂
MW: 331.678 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00245989
CAS Number: 1022610-19-0