5-[4-(Trifluoromethyl)phenyl]-1,3-oxazole
Supplier: Apollo Scientific
Synonyms: 5-(4-(Trifluoromethyl)phenyl)oxazole
PC300556-1G PC300556-5G
APOSPC300556-1GEA 232 EUR
APOSPC300556-1G APOSPC300556-5G
5-[4-(Trifluoromethyl)phenyl]-1,3-oxazole
5-[4-(Trifluoromethyl)phenyl]-1,3-oxazole
Formula: C₁₀H₆F₃NO
MW: 213.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13176792
CAS Number: 87150-14-9