3-Bromo-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine
Supplier: Apollo Scientific
Warning
PC300558-1G PC300558-5G PC300558-250MG
APOSPC300558-1GEA 145 EUR
APOSPC300558-1G APOSPC300558-5G APOSPC300558-250MG
3-Bromo-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine
3-Bromo-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine
Formula: C₈H₃BrClF₃N₂
MW: 299.475 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07021357
CAS Number: 900019-65-0