Ethyl 3-bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Supplier: Apollo Scientific
Warning
PC300576-1G PC300576-250MG
APOSPC300576-1GEA 335 EUR
APOSPC300576-1G APOSPC300576-250MG
Ethyl 3-bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Ethyl 3-bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₁H₈BrF₃N₂O₂
MW: 337.093 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17168280
CAS Number: 1355171-40-2