5-(Trifluoromethyl)-1-benzothiophene-2-carbohydrazide
Supplier: Apollo Scientific
Warning
PC300591-1G PC300591-250MG
APOSPC300591-1GEA 406 EUR
APOSPC300591-1G APOSPC300591-250MG
5-(Trifluoromethyl)-1-benzothiophene-2-carbohydrazide
5-(Trifluoromethyl)-1-benzothiophene-2-carbohydrazide
Formula: C₁₀H₇F₃N₂OS
MW: 260.236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12032266
CAS Number: 1171927-43-7