2-(2-Fluorophenoxy)benzaldehyde
Supplier: Apollo Scientific
PC300604-1G PC300604-5G
APOSPC300604-1GEA 186 EUR
APOSPC300604-1G APOSPC300604-5G
2-(2-Fluorophenoxy)benzaldehyde
2-(2-Fluorophenoxy)benzaldehyde
Formula: C₁₃H₉FO₂
MW: 216.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04108480
CAS Number: 82097-82-3