4-(2-Fluorophenoxy)benzaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-4'-formyldiphenyl ether
PC300611-1G PC300611-5G
APOSPC300611-1GEA 186 EUR
APOSPC300611-1G APOSPC300611-5G
4-(2-Fluorophenoxy)benzaldehyde
4-(2-Fluorophenoxy)benzaldehyde
Formula: C₁₃H₉FO₂
MW: 216.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09026272
CAS Number: 936343-96-3