[4-(2-Fluorophenoxy)phenyl]methanol
Supplier: Apollo Scientific
PC300612-1G PC300612-5G
APOSPC300612-1GEA 248 EUR
APOSPC300612-1G APOSPC300612-5G
[4-(2-Fluorophenoxy)phenyl]methanol
[4-(2-Fluorophenoxy)phenyl]methanol
Formula: C₁₃H₁₁FO₂
MW: 218.224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11190395
CAS Number: 1038966-53-8