N-[4-Bromo-2-(trifluoromethoxy)phenyl]thiourea
Supplier: Apollo Scientific
PC300637-1G PC300637-5G
APOSPC300637-1GEA 149 EUR
APOSPC300637-1G APOSPC300637-5G
N-[4-Bromo-2-(trifluoromethoxy)phenyl]thiourea
N-[4-Bromo-2-(trifluoromethoxy)phenyl]thiourea
Formula: C₈H₆BrF₃N₂OS
MW: 315.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11109537
CAS Number: 1227955-26-1