1-(2-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}phenyl)ethanone
Supplier: Apollo Scientific
PC300645-1G PC300645-10G PC300645-5G
APOSPC300645-1GEA 193 EUR
APOSPC300645-1G APOSPC300645-10G APOSPC300645-5G
1-(2-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}phenyl)ethanone
1-(2-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}phenyl)ethanone
Formula: C₁₃H₉F₃N₂O₂
MW: 282.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00115019
CAS Number: 874782-04-4