N-[4-Bromo-2-(trifluoromethoxy)phenyl]-N'-(2-hydroxyethyl)thiourea
Supplier: Apollo Scientific
PC300685-1G PC300685-5G PC300685-10G
APOSPC300685-1GEA 173 EUR
APOSPC300685-1G APOSPC300685-5G APOSPC300685-10G
N-[4-Bromo-2-(trifluoromethoxy)phenyl]-N'-(2-hydroxyethyl)thiourea
N-[4-Bromo-2-(trifluoromethoxy)phenyl]-N'-(2-hydroxyethyl)thiourea
Formula: C₁₀H₁₀BrF₃N₂O₂S
MW: 359.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18074544
CAS Number: 1427460-44-3