N-[4-Bromo-2-(trifluoromethoxy)phenyl]-4,5-dihydro-1,3-thiazol-2-amine
Supplier: Apollo Scientific
PC300686-1G PC300686-5G
APOSPC300686-1GEA 198 EUR
APOSPC300686-1G APOSPC300686-5G
N-[4-Bromo-2-(trifluoromethoxy)phenyl]-4,5-dihydro-1,3-thiazol-2-amine
N-[4-Bromo-2-(trifluoromethoxy)phenyl]-4,5-dihydro-1,3-thiazol-2-amine
Formula: C₁₀H₈BrF₃N₂OS
MW: 341.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18074545
CAS Number: 1427460-45-4