5-(4-Fluorophenyl)thieno[2,3-d]pyrimidin-4(3H)-one
Supplier: Apollo Scientific
Warning
PC300688-1G PC300688-5G
APOSPC300688-1GEA 198 EUR
APOSPC300688-1G APOSPC300688-5G
5-(4-Fluorophenyl)thieno[2,3-d]pyrimidin-4(3H)-one
5-(4-Fluorophenyl)thieno[2,3-d]pyrimidin-4(3H)-one
Formula: C₁₂H₇FN₂OS
Melting Pt: 190…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00297812
CAS Number: 35978-37-1