2-[2-(Trifluoromethoxy)phenyl]-1,3,4-oxadiazole
Supplier: Apollo Scientific
PC300697-1G PC300697-5G PC300697-10G
APOSPC300697-1GEA 125 EUR
APOSPC300697-1G APOSPC300697-5G APOSPC300697-10G
2-[2-(Trifluoromethoxy)phenyl]-1,3,4-oxadiazole
2-[2-(Trifluoromethoxy)phenyl]-1,3,4-oxadiazole
Formula: C₉H₅F₃N₂O₂
MW: 230.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20441491
CAS Number: 1427460-77-2