2-Bromo-1-(2,2-diethoxyethoxy)-4-fluorobenzene
Supplier: Apollo Scientific
PC300752-10G PC300752-1G PC300752-5G
APOSPC300752-10GEA 864 EUR
APOSPC300752-10G APOSPC300752-1G APOSPC300752-5G
2-Bromo-1-(2,2-diethoxyethoxy)-4-fluorobenzene
2-Bromo-1-(2,2-diethoxyethoxy)-4-fluorobenzene
Formula: C₁₂H₁₆BrFO₃
MW: 307.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14584680
CAS Number: 253429-18-4