8-Bromo-6-fluoroimidazo[1,2-a]pyridine
Supplier: Apollo Scientific
Warning
PC300754-1G PC300754-250MG
APOSPC300754-1GEA 166 EUR
APOSPC300754-1G APOSPC300754-250MG
8-Bromo-6-fluoroimidazo[1,2-a]pyridine
8-Bromo-6-fluoroimidazo[1,2-a]pyridine
Formula: C₇H₄BrFN₂
MW: 215.022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20923664
CAS Number: 1368664-08-7