4-Chloro-N-[2-piperidin-1-yl-5-(trifluoromethyl)phenyl]butanamide
Supplier: Apollo Scientific
PC300758-1G PC300758-5G
APOSPC300758-1GEA 199 EUR
APOSPC300758-1G APOSPC300758-5G
4-Chloro-N-[2-piperidin-1-yl-5-(trifluoromethyl)phenyl]butanamide
4-Chloro-N-[2-piperidin-1-yl-5-(trifluoromethyl)phenyl]butanamide
Formula: C₁₆H₂₀ClF₃N₂O
MW: 348.791 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27995626
CAS Number: 1858255-70-5