Methyl 2,3,5-trifluoro-4-methoxybenzoate
Supplier: Apollo Scientific
Warning
PC300760-1G PC300760-5G
APOSPC300760-1GEA 364 EUR
APOSPC300760-1G APOSPC300760-5G
Methyl 2,3,5-trifluoro-4-methoxybenzoate
Methyl 2,3,5-trifluoro-4-methoxybenzoate
Formula: C₉H₇F₃O₃
MW: 220.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16877111
CAS Number: 1656263-44-3