Methyl 2,5-difluoro-4-methoxybenzoate
Supplier: Apollo Scientific
PC300785-1G PC300785-5G PC300785-250MG
APOSPC300785-1GEA 287 EUR
APOSPC300785-1G APOSPC300785-5G APOSPC300785-250MG
Methyl 2,5-difluoro-4-methoxybenzoate
Methyl 2,5-difluoro-4-methoxybenzoate
Formula: C₉H₈F₂O₃
MW: 202.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399129
CAS Number: 1261830-21-0