3-Chloro-4,4,4-trifluoro-2-phenylbut-2-enal
Supplier: Apollo Scientific
PC300800-1G PC300800-10G PC300800-5G
APOSPC300800-1GEA 223 EUR
APOSPC300800-1G APOSPC300800-10G APOSPC300800-5G
3-Chloro-4,4,4-trifluoro-2-phenylbut-2-enal
3-Chloro-4,4,4-trifluoro-2-phenylbut-2-enal
Formula: C₁₀H₆ClF₃O
MW: 234.602 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972765
CAS Number: 119197-24-9