N-(4-Chlorophenyl)-N'-(4-fluorophenyl)urea
Supplier: Apollo Scientific
PC300821-1G PC300821-5G
APOSPC300821-1GEA 147 EUR
APOSPC300821-1G APOSPC300821-5G
N-(4-Chlorophenyl)-N'-(4-fluorophenyl)urea
N-(4-Chlorophenyl)-N'-(4-fluorophenyl)urea
Formula: C₁₃H₁₀ClFN₂O
MW: 264.683 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00018540
CAS Number: 404-51-3