5-Hydroxy-5-(trifluoromethyl)-4,5-dihydro-1H-pyrazole-1-carbothioamide
Supplier: Apollo Scientific
PC300847-1G PC300847-5G PC300847-10G
APOSPC300847-1GEA 161 EUR
APOSPC300847-1G APOSPC300847-5G APOSPC300847-10G
5-Hydroxy-5-(trifluoromethyl)-4,5-dihydro-1H-pyrazole-1-carbothioamide
5-Hydroxy-5-(trifluoromethyl)-4,5-dihydro-1H-pyrazole-1-carbothioamide
Formula: C₅H₆F₃N₃OS
MW: 213.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03150934
CAS Number: 219986-56-8