N,N-Diisopropyl-6-(trifluoromethyl)nicotinamide
Supplier: Apollo Scientific
PC300861-1G PC300861-5G
APOSPC300861-1GEA 183 EUR
APOSPC300861-1G APOSPC300861-5G
N,N-Diisopropyl-6-(trifluoromethyl)nicotinamide
N,N-Diisopropyl-6-(trifluoromethyl)nicotinamide
Formula: C₁₃H₁₇F₃N₂O
MW: 274.282 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27939889
CAS Number: 765298-12-2