Methyl 4-(difluoromethoxy)benzoate
Supplier: Apollo Scientific
PC300977-10G PC300977-1G PC300977-5G
APOSPC300977-10GEA 902 EUR
APOSPC300977-10G APOSPC300977-1G APOSPC300977-5G
Methyl 4-(difluoromethoxy)benzoate
Methyl 4-(difluoromethoxy)benzoate
Formula: C₉H₈F₂O₃
MW: 202.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04967300
CAS Number: 97914-54-0