6-Bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Supplier: Apollo Scientific
Warning
PC300987-1G PC300987-250MG
APOSPC300987-1GEA 620 EUR
APOSPC300987-1G APOSPC300987-250MG
6-Bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
6-Bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Formula: C₉H₄BrF₃N₂O₂
MW: 309.039 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687630
CAS Number: 1121051-31-7