Ethyl 1-(4-bromo-2-fluorophenyl)-5-oxopyrrolidine-3-carboxylate
Supplier: Apollo Scientific
PC300997-1G PC300997-250MG
APOSPC300997-1GEA 339 EUR
APOSPC300997-1G APOSPC300997-250MG
Ethyl 1-(4-bromo-2-fluorophenyl)-5-oxopyrrolidine-3-carboxylate
Ethyl 1-(4-bromo-2-fluorophenyl)-5-oxopyrrolidine-3-carboxylate
Formula: C₁₃H₁₃BrFNO₃
Storage Temperature: Ambient
MDL Number: MFCD16630891
CAS Number: 1239731-93-1