4-(8-Fluoroimidazo[1,2-a]pyridin-2-yl)benzonitrile
Supplier: Apollo Scientific
Warning
PC301014-1G PC301014-250MG PC301014-5G
APOSPC301014-1GEA 60.9 EUR
APOSPC301014-1G APOSPC301014-250MG APOSPC301014-5G
4-(8-Fluoroimidazo[1,2-a]pyridin-2-yl)benzonitrile
4-(8-Fluoroimidazo[1,2-a]pyridin-2-yl)benzonitrile
Formula: C₁₄H₈FN₃
MW: 237.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32066745
CAS Number: 2366994-15-0