2-(Difluoromethyl)imidazo[1,2-a]pyridine-3-carbohydrazide
Supplier: Apollo Scientific
PC301030-1G PC301030-250MG
APOSPC301030-1GEA 464 EUR
APOSPC301030-1G APOSPC301030-250MG
2-(Difluoromethyl)imidazo[1,2-a]pyridine-3-carbohydrazide
2-(Difluoromethyl)imidazo[1,2-a]pyridine-3-carbohydrazide
Formula: C₉H₈F₂N₄O
MW: 226.183 g/mol
Melting Pt: 151…153 °C
Storage Temperature: Ambient
CAS Number: 2451256-41-8