Ethyl 6-chloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Supplier: Apollo Scientific
PC301032-1G PC301032-250MG
APOSPC301032-1GEA 522 EUR
APOSPC301032-1G APOSPC301032-250MG
Ethyl 6-chloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Ethyl 6-chloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Formula: C₁₁H₉ClF₂N₂O₂
MW: 274.651 g/mol
Melting Pt: 103…106 °C
Storage Temperature: Ambient
CAS Number: 2451256-51-0