2-(Difluoromethyl)-6-fluoroimidazo[1,2-a]pyridine-3-carboxylic acid
Supplier: Apollo Scientific
PC301037-1G PC301037-250MG
APOSPC301037-1GEA 522 EUR
APOSPC301037-1G APOSPC301037-250MG
2-(Difluoromethyl)-6-fluoroimidazo[1,2-a]pyridine-3-carboxylic acid
2-(Difluoromethyl)-6-fluoroimidazo[1,2-a]pyridine-3-carboxylic acid
Formula: C₉H₅F₃N₂O₂
MW: 230.143 g/mol
Melting Pt: 214…215 °C
Storage Temperature: Ambient
MDL Number: MFCD32689366
CAS Number: 2168347-64-4