Ethyl 8-bromo-2-(difluoromethyl)-6-methylimidazo[1,2-a]pyridine-3-carboxylate
Supplier: Apollo Scientific
PC301038-1G PC301038-250MG
APOSPC301038-1GEA 522 EUR
APOSPC301038-1G APOSPC301038-250MG
Ethyl 8-bromo-2-(difluoromethyl)-6-methylimidazo[1,2-a]pyridine-3-carboxylate
Ethyl 8-bromo-2-(difluoromethyl)-6-methylimidazo[1,2-a]pyridine-3-carboxylate
Formula: C₁₂H₁₁BrF₂N₂O₂
MW: 333.129 g/mol
Melting Pt: 128…131 °C
Storage Temperature: Ambient
CAS Number: 2451256-44-1